Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Wayson Stain

Catalog No. p-3201195
Click to view available options
Quantity:
100 mL
2 x 100 mL

The Wayson Stain is for identification of Yersinia Pestis.

  • Contains methylene blue, basic fuchsin, phenol, and ethanol
  • Bipolar staining is characteristic and is best visualized with Wayson Stain
  • Yersinia Pestis organisms stain dark blue, leukocytes stain light blue or purple, and the background stains a light blue
  • Wayson Stain
  • For identification of Yersinia pestis, in-vitro diagnostic use

Specifications

Type Wayson Stain
Quantity 100 mL
Includes Methylene blue, basic fuchsin, phenol, ethanol
Format Bottle
For Use With (Application) Identification of Yersinia pestis, in-vitro diagnostic use
Stain Type Wayson

For in vitro diagnostic use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.