Learn More
ENG Scientific WBC Diluting Fluid
Catalog No. ES5401
Change view
Click to view available options
Quantity:
1 x 100 mL
2 x 100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES5401 1 x 100 mL
ES5400 2 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ES5401 Supplier ENG Scientific Supplier No. 5401
Add to Cart
Add to Cart
WBC Diluting Fluid is a modified solution which hemolyzes RBC's and intensifies the cellular outline of WBC's.
  • Contents: Glacial Acetic Acid
Specifications
Quantity 1 x 100 mL
For Use With (Application) Hemolyze RBCs and intensify cellular outline of WBCs in blood sample
Product Type WBC Diluting Fluid
For in vitro diagnostic use only.