Learn More
ENG Scientific Wright Quick Stain Kits
Description
- Less time in Solution III provides a more alkaline (blue) stain
- Increase time for a more acidic (red) stain
Specifications
Specifications
| Quantity | 3 x 3800 mL (1 gal.) |
| For Use With (Application) | For excellent staining results in one min. |
| Includes | Solution I Wright Stain, Solution II Eosin, Solution III Buffer |
| Packaging | Bottles |
| Product Type | Stain Kit |
For in vitro diagnostic use only.