Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENG Scientific Wright Quick Stain Kits

Catalog No. p-3274918
Change view
Click to view available options
Quantity:
3 x 3800 mL (1 gal.)
3 x 475 mL
Catalog No. Quantity
ES4000 3 x 475 mL
ES4010 3 x 3800 mL (1 gal.)
2 options
This item is not returnable. View return policy

Catalog No. ES4000

Supplier: ENG Scientific 4000

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For excellent staining results in one minute without precipitate. Alcoholic quick stain technique. No fixation necessary. For Quick Stain Kit (Wright or Wright Giemsa).

  • Less time in Solution III provides a more alkaline (blue) stain
  • Increase time for a more acidic (red) stain

Specifications

Quantity 3 x 475 mL
For Use With (Application) For excellent staining results in one min.
Includes Solution I Wright Stain, Solution II Eosin, Solution III Buffer
Packaging Complete Kit (Bottles)

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.