Learn More
ENG Scientific Solution I - Wright Stain
Catalog No. ES4200
Change view
Click to view available options
Quantity:
1 x 3800 mL
Catalog No. Quantity
ES4200 1 x 3800 mL
1 options
Catalog No. ES4200 Supplier ENG Scientific Supplier No. 4200
Add to Cart
Add to Cart
Solution I - Wright Stain for excellent staining results in one minute without precipitate.
  • Alcoholic quick stain technique
  • No fixation necessary
  • For Quick Stain Kit (Wright or Wright Giemsa)
Specifications
Quantity 1 x 3800 mL
For Use With (Application) For Alcoholic quick stain technique, in-vitro diagnostic use only
Product Type Stain
For in vitro diagnostic use only.