Learn More
ENG Scientific Ziehl-Neelsen Stain Kit
Catalog No. ES7000
Change view
Click to view available options
Quantity:
3 x 250 mL
Catalog No. Quantity
ES7000 3 x 250 mL
1 options
Catalog No. ES7000 Supplier ENG Scientific Supplier No. 7000
Add to Cart
Add to Cart
Acid-fast organisms stain red. Other organisms and material stain blue. Use with Ziehl-Neelsen Stain Kit. Kit includes 250 mL each: Solution I - Carbol Fuchsin, Solution II - Acid Alcohol, and Solution III - Loeffler's Methylene Blue.
  • Ziehl Neelsen Stain Kit
  • Includes 250 mL Solution I - Carbol Fuchsin, 250 mL Solution II - Acid Alcohol, 250 mL Solution III - Loeffler's Methylene Blue
  • For Acid fast bacteria
  • 1 Kit
Specifications
Quantity 3 x 250 mL
Includes 250 mL Solution I - carbol fuchsin, 250 mL Solution II - acid alcohol, 250 mL Solution III - Loeffler's methylene blue
For Use With (Application) Acid fast bacteria
Stain Type Ziehl-Neelsen
Volume (Metric) 250 mL
Product Type Ziehl Neelsen Stain Kit
Packaging Complete Kit (Bottles)
For in vitro diagnostic use only.