Learn More
Terumo Engauge Hypodermic Syringe without Needle
Catalog No. 22290044 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Needle Gauge:
No Needle Included
Catalog No. Needle Gauge
22-290-044 No Needle Included
1 options
This item is not returnable. View return policy
Catalog No. 22-290-044 Supplier Terumo Supplier No. SENGSS01L
This item is not returnable. View return policy
Engauge 1 cc Hypodermic Syringe with Low Dead Space
  • Low dead space
  • Luer Lock
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polypropylene (Plunger), Polypropylene (Hub)
Needle Gauge No Needle Included
Latex Free Yes
Disposable Single-use
Volume (Metric) 1 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Lock
Graduated Yes
Sterility Sterile