Learn More
Accuflow Systems Engel Servo Motor, Synchronous, 48 VDC, 485 W
Catalog No. NC3849326
Encompass_Preferred
Synchronous servo motor designed for industrial automation and pump drive applications. Typical specifications include 48 VDC supply and approximately 485 W output. Ingress protection rating varies by configuration and source reporting.
  • Suitable for drive applications in fluid transfer and automation
  • Rated for 48 VDC operation
  • Approximately 485 W output power
  • Sealed enclosure, IP rating varies by build
  • Black painted finish reported in supplier documentation
Catalog No. NC3849326 Supplier Accuflow Systems Supplier No. HBI37904A
Synchronous servo motor designed for industrial automation and pump drive applications. Typical specifications include 48 VDC supply and approximately 485 W output. Ingress protection rating varies by configuration and source reporting.
  • Suitable for drive applications in fluid transfer and automation
  • Rated for 48 VDC operation
  • Approximately 485 W output power
  • Sealed enclosure, IP rating varies by build
  • Black painted finish reported in supplier documentation