Learn More
Medchemexpress LLC Engeletin | 572-31-6 | 434.39 | 1 MG
SDP
Catalog No. 5000389993
Encompass_Preferred
Engeletin is a flavanonol glycoside isolated from Smilax glabra Roxb. It inhibits NF-κB signaling-pathway activation and has anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects.
  • Isolated from Smilax glabra Roxb.
  • Inhibits NF-κB signaling-pathway activation.
  • Possesses anti-inflammatory effects.
  • Possesses analgesic effects.
  • Possesses diuresis effects.
  • Possesses detumescence effects.
  • Possesses antibiosis effects.
  • Markedly reduces LPS-increased myeloperoxidase activity in mice.
  • Decreases the production of inflammatory mediators (iNOS and COX-2).
  • Suppresses the expression of TLR4-signaling downstream molecules such as MyD88, IRAK1, TRAF6, and TAK1 proteins.
Catalog No. 50-003-89993 Supplier Medchemexpress LLC Supplier No. HYN04361MG
Engeletin is a flavanonol glycoside isolated from Smilax glabra Roxb. It inhibits NF-κB signaling-pathway activation and has anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects.
  • Isolated from Smilax glabra Roxb.
  • Inhibits NF-κB signaling-pathway activation.
  • Possesses anti-inflammatory effects.
  • Possesses analgesic effects.
  • Possesses diuresis effects.
  • Possesses detumescence effects.
  • Possesses antibiosis effects.
  • Markedly reduces LPS-increased myeloperoxidase activity in mice.
  • Decreases the production of inflammatory mediators (iNOS and COX-2).
  • Suppresses the expression of TLR4-signaling downstream molecules such as MyD88, IRAK1, TRAF6, and TAK1 proteins.