Learn More
Medchemexpress LLC Engeletin | 572-31-6 | 434.39 | 100 MG
SDP
Catalog No. 5000389125
Encompass_Preferred
Engeletin is a flavanonol glycoside isolated from Smilax glabra Roxb. It inhibits NF-κB signaling-pathway activation.
  • Possesses anti-inflammatory effects.
  • Possesses analgesic effects.
  • Possesses diuresis effects.
  • Possesses detumescence effects.
  • Possesses antibiosis effects.
Catalog No. 50-003-89125 Supplier Medchemexpress LLC Supplier No. HYN0436100MG
Engeletin is a flavanonol glycoside isolated from Smilax glabra Roxb. It inhibits NF-κB signaling-pathway activation.
  • Possesses anti-inflammatory effects.
  • Possesses analgesic effects.
  • Possesses diuresis effects.
  • Possesses detumescence effects.
  • Possesses antibiosis effects.