Learn More
Medchemexpress LLC Engeletin | 572-31-6 | 434.39 | 25 MG
SDP
Catalog No. 5000389960
Encompass_Preferred
Engeletin is a flavanonol glycoside isolated from *Smilax glabra Roxb.* It acts as an inhibitor of NF-κB signaling-pathway activation and exhibits various biological effects, including anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis properties. For research use only.
  • Inhibits NF-κB signaling-pathway activation
  • Possesses anti-inflammatory effects
  • Offers analgesic properties
  • Exhibits diuresis and detumescence effects
  • Has antibiosis effects
Catalog No. 50-003-89960 Supplier Medchemexpress LLC Supplier No. 25MGHYN0436
Engeletin is a flavanonol glycoside isolated from *Smilax glabra Roxb.* It acts as an inhibitor of NF-κB signaling-pathway activation and exhibits various biological effects, including anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis properties. For research use only.
  • Inhibits NF-κB signaling-pathway activation
  • Possesses anti-inflammatory effects
  • Offers analgesic properties
  • Exhibits diuresis and detumescence effects
  • Has antibiosis effects