Learn More
Medchemexpress LLC Engeletin | 572-31-6 | 99.9% | 434.39 g/mol | C21H22O10 | 10 MG
SDP
Catalog No. 5000295646
Encompass_Preferred
Engeletin is a flavanonol glycoside reported to inhibit NF-κB signaling and to exhibit anti-inflammatory, analgesic, diuretic, and antibacterial effects. It is intended for research use only and has high solubility in DMSO but is essentially insoluble in water.
  • Inhibits NF-κB signaling pathway.
  • Shows anti-inflammatory and analgesic activity.
  • High solubility in DMSO, insoluble in water.
  • High purity (99.9%).
  • Provided as a small-mass research reagent suitable for in vitro studies.
Catalog No. 50-002-95646 Supplier Medchemexpress LLC Supplier No. HYN043610MG
Engeletin is a flavanonol glycoside reported to inhibit NF-κB signaling and to exhibit anti-inflammatory, analgesic, diuretic, and antibacterial effects. It is intended for research use only and has high solubility in DMSO but is essentially insoluble in water.
  • Inhibits NF-κB signaling pathway.
  • Shows anti-inflammatory and analgesic activity.
  • High solubility in DMSO, insoluble in water.
  • High purity (99.9%).
  • Provided as a small-mass research reagent suitable for in vitro studies.