Learn More
Medchemexpress LLC HY-N0436 5mg Medchemexpress, Engeletin CAS:572-31-6 Purity:>98%
SDP
Catalog No. 501933570
Encompass_Preferred
Medchemexpress, HY-N0436 5mg Engeletin CAS:572-31-6 Engeletin is a flavanonol glycoside isolated from hymenaea martiana, inhibits NF-κB signaling-pathway activation, and possesses anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3570 Supplier Medchemexpress LLC Supplier No. HYN04365MG
Medchemexpress, HY-N0436 5mg Engeletin CAS:572-31-6 Engeletin is a flavanonol glycoside isolated from hymenaea martiana, inhibits NF-κB signaling-pathway activation, and possesses anti-inflammatory, analgesic, diuresis, detumescence, and antibiosis effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.