Learn More
New England Biolabs, Inc. EnGen Cas9 NLS, S. pyogenes – 400 pmol
SDP
Catalog No. NC1194378
Encompass
EnGen Spy Cas9 NLS, is an RNA-guided endonuclease that catalyzes site- specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) ). The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence. EnGen Spy Cas9 NLS contains Simian virus 40 (SV40) T antigen nuclear localization sequence (NLS) on the N- and C- termini of the protein.
Catalog No. NC1194378 Supplier New England Biolabs, Inc. Supplier No. M0646T
EnGen Spy Cas9 NLS, is an RNA-guided endonuclease that catalyzes site- specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) ). The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence. EnGen Spy Cas9 NLS contains Simian virus 40 (SV40) T antigen nuclear localization sequence (NLS) on the N- and C- termini of the protein.