Learn More
New England Biolabs, Inc. EnGen Spy Cas9 HF1 - 2500 pmol
SDP
Catalog No. 502336251
Encompass
EnGen Spy Cas9 HF1 is a high-fidelity, quadruple substitution (N497A/R661A/Q695A/Q926A) variant of EnGen Spy Cas9 NLS from Streptococcus pyogenes with reduced non-specific DNA cleavage. Spy Cas9 is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The single guide RNA (sgRNA) targets Cas9 to the region immediately upstream of a 5'-NGG-3' protospacer adjacent motif (PAM) producing a double stranded break 3 bases upstream of the PAM. EnGen Spy Cas9 HF1 contains Simian virus 40 (SV40) T antigen nuclear localization sequence (NLS) on the N- and C-termini of the protein.
Catalog No. 50-233-6251 Supplier New England Biolabs, Inc. Supplier No. M0667M
EnGen Spy Cas9 HF1 is a high-fidelity, quadruple substitution (N497A/R661A/Q695A/Q926A) variant of EnGen Spy Cas9 NLS from Streptococcus pyogenes with reduced non-specific DNA cleavage. Spy Cas9 is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The single guide RNA (sgRNA) targets Cas9 to the region immediately upstream of a 5'-NGG-3' protospacer adjacent motif (PAM) producing a double stranded break 3 bases upstream of the PAM. EnGen Spy Cas9 HF1 contains Simian virus 40 (SV40) T antigen nuclear localization sequence (NLS) on the N- and C-termini of the protein.