Learn More
KLARMANN RULINGS INC ENGLAND FINDER
Catalog No. NC9122344
Encompass_Preferred
England finder; Klarmann; Glass slide marked over the top surface in a way that a referenced position can be directly read relative to the locating edges
Catalog No. NC9122344 Supplier KLARMANN RULINGS INC Supplier No. KR834
England finder; Klarmann; Glass slide marked over the top surface in a way that a referenced position can be directly read relative to the locating edges