Learn More
Electron Microscopy Sciences English Scale Only for 10x Magnifier
SDP
Catalog No. 5027058
Encompass_Preferred
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Field of view: 28mm, eyepiece: 20mm, scale diameter: 35mm, scale: 20mm; divisions: 0.8", 0.005". Metric & English scale interchangeable. Size: 46mm
Catalog No. 50-270-58 Supplier Electron Microscopy Sciences Supplier No. 68155
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Field of view: 28mm, eyepiece: 20mm, scale diameter: 35mm, scale: 20mm; divisions: 0.8", 0.005". Metric & English scale interchangeable. Size: 46mm