Learn More
Electron Microscopy Sciences English Scale Only for 15x Magnifier
SDP
Supplier:  Electron Microscopy Sciences 68159
Encompass_Preferred
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Field of view: 14mm, eyepiece: 13mm, scale diameter: 26mm, scale: 20mm; divisions: 0.8", 0.005". Metric & English interchangeable. Size: 35mm
Catalog No. 50-270-62