Learn More
Electron Microscopy Sciences English Scale for 7x Magnifier
SDP
Supplier:  Electron Microscopy Sciences 68151
Encompass_Preferred
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Lens: 19mm, eyepiece: 18mm, scale diameter: 26mm measuring scale: 20mm, divisions: 0.8", divisions: 0.005". Metric & English scale interchangeable. Size: 36mm (d)
Catalog No. 50-270-60