Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502764069
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R941 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM. The tool has a bright finish for enhanced workpiece visibility.
Catalog No. 50-276-4069 Supplier Grainger Supplier No. 16R940
The Micro 100 Engraving Tool Single End 16R941 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM. The tool has a bright finish for enhanced workpiece visibility.