Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502763871
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R928 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work and making fine lines. The single end cutting edge allows for precision control. Made in the USA from high-speed steel, this engraving tool offers good wear resistance and hardness for engraving applications.
Catalog No. 50-276-3871 Supplier Grainger Supplier No. 16R928
The Micro 100 Engraving Tool Single End 16R928 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work and making fine lines. The single end cutting edge allows for precision control. Made in the USA from high-speed steel, this engraving tool offers good wear resistance and hardness for engraving applications.