Learn More
Grainger Micro 100 Engraving Cutter, Solid Carbide, 60°, 1/4 in. Shank, 1 ea
Catalog No. 502763935
Encompass_Preferred
This solid carbide engraving tool is designed for high-precision engraving and part identification applications. Featuring a single-end, tipped-off conical geometry, it provides reliable performance in a variety of materials, with a primary application focus on steel.
  • Solid carbide construction for durability
  • 60° included angle for precise marking
  • 0.25 in. shank diameter
  • 0.004 in. point offset with a ±0.001 in. tolerance
  • Uncoated finish
  • Right-hand cutting direction
  • Designed for finishing applications
Catalog No. 50-276-3935 Supplier Grainger Supplier No. 16R925
This solid carbide engraving tool is designed for high-precision engraving and part identification applications. Featuring a single-end, tipped-off conical geometry, it provides reliable performance in a variety of materials, with a primary application focus on steel.
  • Solid carbide construction for durability
  • 60° included angle for precise marking
  • 0.25 in. shank diameter
  • 0.004 in. point offset with a ±0.001 in. tolerance
  • Uncoated finish
  • Right-hand cutting direction
  • Designed for finishing applications