Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502764550
Encompass_Preferred
The 16R937 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-conductive, non-corrosive agent that leaves no residue and is safe for use around sensitive electronics. The extinguisher has a chrome-plated steel cylinder with an enameled aluminum valve body and discharge hose. It comes equipped with a wall hook and mounting bracket for easy installation. This compact and lightweight fire extinguisher provides fast and effective fire suppression.
Catalog No. 50-276-4550 Supplier Grainger Supplier No. 16R937
The 16R937 is a 3M Novec 1230 Clean Agent Fire Extinguisher. It is UL rated 2-B:C and contains 5 lbs of Novec 1230 fluid. This clean agent extinguisher is intended for Class A, B, and C fires. It uses a non-conductive, non-corrosive agent that leaves no residue and is safe for use around sensitive electronics. The extinguisher has a chrome-plated steel cylinder with an enameled aluminum valve body and discharge hose. It comes equipped with a wall hook and mounting bracket for easy installation. This compact and lightweight fire extinguisher provides fast and effective fire suppression.