Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502763731
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R998 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for fine detail engraving work in applications like engraving nameplates, panels, tags and more. The single end cutting edge allows for engraving in tight spaces.
Catalog No. 50-276-3731 Supplier Grainger Supplier No. 16R998
The Micro 100 Engraving Tool Single End 16R998 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for fine detail engraving work in applications like engraving nameplates, panels, tags and more. The single end cutting edge allows for engraving in tight spaces.