Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502764552
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R941 is a single-ended carbide engraving tool with a 0.010" tip diameter and 0.250" shank diameter. It has a 30 degree included angle and is made of solid carbide for engraving in steel, aluminum, and other non-ferrous metals. The overall length is 3" and it has a bright finish. This is a high quality precision engraving tool suitable for detailed engraving work in metals.
Catalog No. 50-276-4552 Supplier Grainger Supplier No. 16R941
The Micro 100 Engraving Tool Single End 16R941 is a single-ended carbide engraving tool with a 0.010" tip diameter and 0.250" shank diameter. It has a 30 degree included angle and is made of solid carbide for engraving in steel, aluminum, and other non-ferrous metals. The overall length is 3" and it has a bright finish. This is a high quality precision engraving tool suitable for detailed engraving work in metals.