Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502764637
Encompass_Preferred
The Grainger 16T004 engraving tool is a carbide tipped engraver with a 1/8 inch shank diameter and a 60 degree included angle. It has a 0.004 inch tip width and is made of solid carbide for durability and long tool life. The engraver can be used for detail engraving work in metals, plastics, wood and other materials. With its micro grain carbide construction, it provides a sharp cutting edge for clean, crisp detail engraving. The 1/8 inch shank allows it to fit in standard engraving machines. Overall, this is a high quality solid carbide engraving tool well-suited for detailed engraving work.
Catalog No. 50-276-4637 Supplier Grainger Supplier No. 16T004
The Grainger 16T004 engraving tool is a carbide tipped engraver with a 1/8 inch shank diameter and a 60 degree included angle. It has a 0.004 inch tip width and is made of solid carbide for durability and long tool life. The engraver can be used for detail engraving work in metals, plastics, wood and other materials. With its micro grain carbide construction, it provides a sharp cutting edge for clean, crisp detail engraving. The 1/8 inch shank allows it to fit in standard engraving machines. Overall, this is a high quality solid carbide engraving tool well-suited for detailed engraving work.