Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502766709
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R876 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is made of M2 high speed steel for wear resistance and has a bright finish. It can machine a variety of materials including steel, aluminum, brass, copper, plastics and wood. The single end cutting edge allows for detail work in corners and confined areas.
Catalog No. 50-276-6709 Supplier Grainger Supplier No. 16R876
The Micro 100 Engraving Tool Single End 16R876 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is made of M2 high speed steel for wear resistance and has a bright finish. It can machine a variety of materials including steel, aluminum, brass, copper, plastics and wood. The single end cutting edge allows for detail work in corners and confined areas.