Learn More
Grainger Engraving Tool 1/2 L of Cut Carbide
Catalog No. 502767412
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R876 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its small cutting edge. The tool has a 3 inch reach for engraving recessed areas. It is made in the USA from M2 high speed steel for good wear resistance and heat tolerance. The single end cutting edge allows for engraving in tight spaces.
Catalog No. 50-276-7412 Supplier Grainger Supplier No. 16R879
The Micro 100 Engraving Tool Single End 16R876 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its small cutting edge. The tool has a 3 inch reach for engraving recessed areas. It is made in the USA from M2 high speed steel for good wear resistance and heat tolerance. The single end cutting edge allows for engraving in tight spaces.