Learn More
Grainger Engraving Tool 10.00mm L of Cut Carbide
Catalog No. 502763912
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16T022 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a flute length of 1-1/4". The tool features a 30 degree helix angle and a 0.005" tip radius for detailed engraving work in materials like plastics, soft metals, wood, and composites. With its micro grain high-speed steel construction, this engraving tool provides good wear resistance and tool life. The single end cutting edge allows for engraving in tight areas. Overall, this is a versatile hand engraving tool well-suited for detailed engraving work.
Catalog No. 50-276-3912 Supplier Grainger Supplier No. 16T022
The Micro 100 Engraving Tool Single End 16T022 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a flute length of 1-1/4". The tool features a 30 degree helix angle and a 0.005" tip radius for detailed engraving work in materials like plastics, soft metals, wood, and composites. With its micro grain high-speed steel construction, this engraving tool provides good wear resistance and tool life. The single end cutting edge allows for engraving in tight areas. Overall, this is a versatile hand engraving tool well-suited for detailed engraving work.