Learn More
Grainger Engraving Tool 10.00mm L of Cut Carbide
Catalog No. 502763913
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16T025 is a double-ended carbide engraving tool with a 0.016" tip diameter. It has a 1/4" shank diameter and is made of solid carbide for durability. The tool has a 30 degree included angle and is uncoated. It can be used for detail engraving work in materials like steel, aluminum, and plastics. The double-ended design provides two cutting edges for extended tool life. Overall, this Micro 100 engraving tool is well-suited for fine detail engraving work where a sharp, narrow tip is needed.
Catalog No. 50-276-3913 Supplier Grainger Supplier No. 16T025
The Micro 100 Engraving Tool Double End 16T025 is a double-ended carbide engraving tool with a 0.016" tip diameter. It has a 1/4" shank diameter and is made of solid carbide for durability. The tool has a 30 degree included angle and is uncoated. It can be used for detail engraving work in materials like steel, aluminum, and plastics. The double-ended design provides two cutting edges for extended tool life. Overall, this Micro 100 engraving tool is well-suited for fine detail engraving work where a sharp, narrow tip is needed.