Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502763732
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16T033 is a single end engraving tool made by Micro 100. It has a 0.016" tip diameter and 0.250" shank diameter. The overall tool length is 3.00". It is made of solid carbide and has an AlTiN coating. The included angle is 60 degrees. It has a right hand spiral flute. The engraving tool is designed for use on materials like plastics, wood, aluminum and brass. It can be used for detail engraving work in machines like CNC routers and mills. The Micro 100 engraving tool provides good chip clearance and precision results.
Catalog No. 50-276-3732 Supplier Grainger Supplier No. 16T037
The Micro 100 Engraving Tool Single End 16T033 is a single end engraving tool made by Micro 100. It has a 0.016" tip diameter and 0.250" shank diameter. The overall tool length is 3.00". It is made of solid carbide and has an AlTiN coating. The included angle is 60 degrees. It has a right hand spiral flute. The engraving tool is designed for use on materials like plastics, wood, aluminum and brass. It can be used for detail engraving work in machines like CNC routers and mills. The Micro 100 engraving tool provides good chip clearance and precision results.