Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502764632
Encompass_Preferred
The Micro 100 Engraving Tool is a 12mm long carbide engraving cutter with a 0.8mm shank diameter and a 30 degree helix angle. It has a 0.1mm corner radius and is made of solid carbide for wear resistance when engraving into materials like steel, aluminum, and plastics. The Micro 100 engraver features a TiAlN coating for heat and wear resistance. It is designed for light to medium duty engraving applications.
Catalog No. 50-276-4632 Supplier Grainger Supplier No. 16R969
The Micro 100 Engraving Tool is a 12mm long carbide engraving cutter with a 0.8mm shank diameter and a 30 degree helix angle. It has a 0.1mm corner radius and is made of solid carbide for wear resistance when engraving into materials like steel, aluminum, and plastics. The Micro 100 engraver features a TiAlN coating for heat and wear resistance. It is designed for light to medium duty engraving applications.