Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502763951
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16T030 is a double-ended carbide engraving tool with a 1/8" shank diameter. It has a 30 degree included angle and a 0.030" tip radius. The overall length is 2-1/2". It is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. The double-ended design provides two cutting edges for extended tool life. Made in the USA from solid carbide, this high quality engraving tool is rigid and durable for precision engraving results.
Catalog No. 50-276-3951 Supplier Grainger Supplier No. 16T030
The Micro 100 Engraving Tool Double End 16T030 is a double-ended carbide engraving tool with a 1/8" shank diameter. It has a 30 degree included angle and a 0.030" tip radius. The overall length is 2-1/2". It is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. The double-ended design provides two cutting edges for extended tool life. Made in the USA from solid carbide, this high quality engraving tool is rigid and durable for precision engraving results.