Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502763975
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16T034 is a double-ended engraving tool made by Micro 100. It has a 0.016" diameter high speed steel cutter on each end, allowing for detail work in tight spaces. The overall tool length is 3" and it has a 1/4" shank diameter. This engraving tool is designed for use on materials like plastics, wood, aluminum and brass. It can be used for detail engraving work as well as small carving, routing and profiling applications. The double-ended design provides two cutting edges for extended tool life.
Catalog No. 50-276-3975 Supplier Grainger Supplier No. 16T034
The Micro 100 Engraving Tool Double End 16T034 is a double-ended engraving tool made by Micro 100. It has a 0.016" diameter high speed steel cutter on each end, allowing for detail work in tight spaces. The overall tool length is 3" and it has a 1/4" shank diameter. This engraving tool is designed for use on materials like plastics, wood, aluminum and brass. It can be used for detail engraving work as well as small carving, routing and profiling applications. The double-ended design provides two cutting edges for extended tool life.