Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502763872
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R966 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/16 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its sharp pointed tip. The tool has a bright finish for high visibility and comes with a 3/16 inch reduced shank for use in mini die grinders. It is made in the USA by Micro 100, a manufacturer of rotary cutting tools.
Catalog No. 50-276-3872 Supplier Grainger Supplier No. 16R966
The Micro 100 Engraving Tool Single End 16R966 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/16 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its sharp pointed tip. The tool has a bright finish for high visibility and comes with a 3/16 inch reduced shank for use in mini die grinders. It is made in the USA by Micro 100, a manufacturer of rotary cutting tools.