Learn More
Grainger Engraving Tool 12.00mm L of Cut Carbide
Catalog No. 502763877
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16R965 is a double-ended carbide engraving tool designed for engraving, scribing, and deburring in CNC machines. It has a 1/8 inch shank diameter and comes in a set of 2, with one end having a 30 degree chamfer and the other end having a 60 degree chamfer. The overall tool lengths are 1-1/2 inches and 2 inches. This versatile tool provides excellent results in steel, cast iron, aluminum, copper, brass, bronze, plastic and wood. It is rigid and durable for long life. The Micro 100 brand is made in the USA and known for high performance and precision.
Catalog No. 50-276-3877 Supplier Grainger Supplier No. 16R965
The Micro 100 Engraving Tool Double End 16R965 is a double-ended carbide engraving tool designed for engraving, scribing, and deburring in CNC machines. It has a 1/8 inch shank diameter and comes in a set of 2, with one end having a 30 degree chamfer and the other end having a 60 degree chamfer. The overall tool lengths are 1-1/2 inches and 2 inches. This versatile tool provides excellent results in steel, cast iron, aluminum, copper, brass, bronze, plastic and wood. It is rigid and durable for long life. The Micro 100 brand is made in the USA and known for high performance and precision.