Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502765905
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16R852 is a double-ended carbide engraving tool with a 1/8" shank diameter. It has a 60 degree included angle and a 0.010" tip radius. The overall length is 2-1/2". It is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. The double-ended design provides two cutting edges for extended tool life. Made in the USA from solid carbide, this high quality engraving tool is rigid and durable for precision engraving results.
Catalog No. 50-276-5905 Supplier Grainger Supplier No. 16R852
The Micro 100 Engraving Tool Double End 16R852 is a double-ended carbide engraving tool with a 1/8" shank diameter. It has a 60 degree included angle and a 0.010" tip radius. The overall length is 2-1/2". It is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. The double-ended design provides two cutting edges for extended tool life. Made in the USA from solid carbide, this high quality engraving tool is rigid and durable for precision engraving results.