Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502766449
Encompass_Preferred
The Micro 100 Grooving Tool is a carbide insert grooving tool with a 5/16 inch shank diameter. It has a 0.062 inch groove width capacity and can groove up to 0.500 inches deep. The tool holder accepts DGN style inserts and includes one DGNR 331 insert. It is made of solid carbide for strength and rigidity. The grooving tool can be used for grooving, cutoff, and threading applications in steel, stainless steel, cast iron, aluminum and other materials. It has a maximum RPM of 4000. The overall tool length is 5 inches.
Catalog No. 50-276-6449 Supplier Grainger Supplier No. 16R856
The Micro 100 Grooving Tool is a carbide insert grooving tool with a 5/16 inch shank diameter. It has a 0.062 inch groove width capacity and can groove up to 0.500 inches deep. The tool holder accepts DGN style inserts and includes one DGNR 331 insert. It is made of solid carbide for strength and rigidity. The grooving tool can be used for grooving, cutoff, and threading applications in steel, stainless steel, cast iron, aluminum and other materials. It has a maximum RPM of 4000. The overall tool length is 5 inches.