Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502763870
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R917 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point for detail engraving in materials like plastics, soft metals, wood, and fiberglass. It can be used in low horsepower engraving machines and rotary hand engravers. The tool's high-speed steel construction provides wear resistance and heat resistance during use.
Catalog No. 50-276-3870 Supplier Grainger Supplier No. 16R917
The Micro 100 Engraving Tool Single End 16R917 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 3 inches. The tool features a 135 degree point for detail engraving in materials like plastics, soft metals, wood, and fiberglass. It can be used in low horsepower engraving machines and rotary hand engravers. The tool's high-speed steel construction provides wear resistance and heat resistance during use.