Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502767445
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R858 is a high-speed steel engraving cutter with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work in applications like sign making, jewelry engraving, and electronic chassis marking. The single end cutting edge allows for engraving in tight spaces.
Catalog No. 50-276-7445 Supplier Grainger Supplier No. 16R858
The Micro 100 Engraving Tool Single End 16R858 is a high-speed steel engraving cutter with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work in applications like sign making, jewelry engraving, and electronic chassis marking. The single end cutting edge allows for engraving in tight spaces.