Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502763984
Encompass_Preferred
The Grainger 3/8" L of Cut Carbide Engraving Tool is a high-speed steel engraving cutter with a 3/8" length of cut. It has a 1/8" shank diameter and features a 135 degree point angle for detail work in metals, plastics, wood, and composites. This single flute cutter is uncoated and made in the USA. It can be used in low and high speed engraving, routing, and milling applications. The carbide construction provides wear resistance and strength.
Catalog No. 50-276-3984 Supplier Grainger Supplier No. 16R982
The Grainger 3/8" L of Cut Carbide Engraving Tool is a high-speed steel engraving cutter with a 3/8" length of cut. It has a 1/8" shank diameter and features a 135 degree point angle for detail work in metals, plastics, wood, and composites. This single flute cutter is uncoated and made in the USA. It can be used in low and high speed engraving, routing, and milling applications. The carbide construction provides wear resistance and strength.