Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502764549
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R918 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its small cutting edge. The tool has a 3 inch reach for engraving recessed areas. It is made in the USA from M2 high speed steel for good wear resistance and heat tolerance. The 16R918 is a quality engraving tool well suited for detailed hand engraving work.
Catalog No. 50-276-4549 Supplier Grainger Supplier No. 16R918
The Micro 100 Engraving Tool Single End 16R918 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch, an overall length of 3 inches, and a cutting diameter of 1/32 inch. This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can produce fine detailed engravings with its small cutting edge. The tool has a 3 inch reach for engraving recessed areas. It is made in the USA from M2 high speed steel for good wear resistance and heat tolerance. The 16R918 is a quality engraving tool well suited for detailed hand engraving work.