Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502764641
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R984 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point angle for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM in CNC machines. The tool has a bright finish for visibility and is made in the USA by Micro 100.
Catalog No. 50-276-4641 Supplier Grainger Supplier No. 16R984
The Micro 100 Engraving Tool Single End 16R984 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8 inch and an overall length of 4 inches. The tool features a 135 degree point angle for detail engraving in materials like plastics, wood, aluminum and brass. It can be used in low to medium production runs at spindle speeds up to 15,000 RPM in CNC machines. The tool has a bright finish for visibility and is made in the USA by Micro 100.