Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502765022
Encompass_Preferred
The Micro 100 Grooving Tool with a 5/16" shank is a carbide insert grooving tool designed for grooving, cutoff, and parting applications. It has a 0.062" max grooving depth and uses TPG322 style inserts with a 0.031" nose radius. The tool body is made from carbide and has a 5/16" shank diameter. It is capable of grooving steel, stainless steel, cast iron, aluminum, and other materials. The Micro 100 grooving tool allows efficient grooving in a variety of materials while providing good chip control.
Catalog No. 50-276-5022 Supplier Grainger Supplier No. 16R854
The Micro 100 Grooving Tool with a 5/16" shank is a carbide insert grooving tool designed for grooving, cutoff, and parting applications. It has a 0.062" max grooving depth and uses TPG322 style inserts with a 0.031" nose radius. The tool body is made from carbide and has a 5/16" shank diameter. It is capable of grooving steel, stainless steel, cast iron, aluminum, and other materials. The Micro 100 grooving tool allows efficient grooving in a variety of materials while providing good chip control.