Learn More
Grainger Engraving Tool 3/8 L of Cut Carbide
Catalog No. 502763716
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R981 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for fine detail engraving work and marking identification numbers. The single end cutting edge allows for engraving in tight spaces. Made in the USA from M2 high speed steel for good wear resistance and heat tolerance.
Catalog No. 50-276-3716 Supplier Grainger Supplier No. 16R981
The Micro 100 Engraving Tool Single End 16R981 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for fine detail engraving work and marking identification numbers. The single end cutting edge allows for engraving in tight spaces. Made in the USA from M2 high speed steel for good wear resistance and heat tolerance.