Learn More
Grainger Engraving Tool 5.00mm L of Cut Carbide
Catalog No. 502764153
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R888 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work and making fine lines. The single end cutting edge allows for precision control. Made in the USA from M2 high speed steel, this durable engraving tool will retain its sharp cutting edge during extended use.
Catalog No. 50-276-4153 Supplier Grainger Supplier No. 16R888
The Micro 100 Engraving Tool Single End 16R888 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2-1/2", and a cutting diameter of 1/16". This engraving tool is designed for use on materials like plastics, wood, fiberglass and soft non-ferrous metals. It can be used for detailed engraving work and making fine lines. The single end cutting edge allows for precision control. Made in the USA from M2 high speed steel, this durable engraving tool will retain its sharp cutting edge during extended use.