Learn More
Grainger Engraving Tool 5.00mm L of Cut Carbide
Catalog No. 502763804
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R890 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". This engraving tool is made of solid carbide and can machine a variety of materials including steel, aluminum, brass, copper, plastics, and wood. It features a TiAlN coating for increased tool life. The single end cutting edge allows for detail work in tight areas. This is a high quality carbide engraving tool well-suited for detailed engraving work.
Catalog No. 50-276-3804 Supplier Grainger Supplier No. 16R890
The Micro 100 Engraving Tool Single End 16R890 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". This engraving tool is made of solid carbide and can machine a variety of materials including steel, aluminum, brass, copper, plastics, and wood. It features a TiAlN coating for increased tool life. The single end cutting edge allows for detail work in tight areas. This is a high quality carbide engraving tool well-suited for detailed engraving work.