Learn More
Grainger Engraving Tool 5/8 L of Cut Carbide
Catalog No. 502763873
Encompass_Preferred
The Micro 100 Engraving Tool Double End 16R947 is a double-ended carbide engraving tool made in the USA by Micro 100. It has a 1/8" shank diameter and comes in a set of 2, with one end having a 30 degree included angle and the other end having a 60 degree included angle. The overall tool length is 2-1/2". This versatile tool can be used for detail engraving work in most machinable materials.
Catalog No. 50-276-3873 Supplier Grainger Supplier No. 16R947
The Micro 100 Engraving Tool Double End 16R947 is a double-ended carbide engraving tool made in the USA by Micro 100. It has a 1/8" shank diameter and comes in a set of 2, with one end having a 30 degree included angle and the other end having a 60 degree included angle. The overall tool length is 2-1/2". This versatile tool can be used for detail engraving work in most machinable materials.