Learn More
Grainger Engraving Tool 5/8 L of Cut Carbide
Catalog No. 502766649
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R883 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 3", and a cutting diameter of 1/16". This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can be used in low-powered rotary hand engravers. The single end cutting edge allows for detail engraving work. Made of M2 high-speed steel, it maintains a sharp edge at high engraving speeds. The Micro 100 engraving tool is suitable for hobbyists and professionals looking for a quality steel engraving tool for detailed hand engraving work.
Catalog No. 50-276-6649 Supplier Grainger Supplier No. 16R883
The Micro 100 Engraving Tool Single End 16R883 is a high-speed steel engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 3", and a cutting diameter of 1/16". This engraving tool is designed for hand engraving in materials like plastics, soft metals, wood, and fiberglass. It can be used in low-powered rotary hand engravers. The single end cutting edge allows for detail engraving work. Made of M2 high-speed steel, it maintains a sharp edge at high engraving speeds. The Micro 100 engraving tool is suitable for hobbyists and professionals looking for a quality steel engraving tool for detailed hand engraving work.