Learn More
Grainger Engraving Tool 5/8 L of Cut Carbide
Catalog No. 502766701
Encompass_Preferred
The Micro 100 Engraving Tool Single End 16R880 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". This engraving tool is made of solid carbide and can machine a variety of materials including steel, aluminum, brass, copper, plastics, and wood. With its micro grain carbide construction, it provides excellent wear resistance and tool life. The single end cutting edge allows for detail engraving work in tight areas. This is a high precision engraving tool well-suited for detailed machining in applications like engraving, die sinking, cavity sinking, and micro machining.
Catalog No. 50-276-6701 Supplier Grainger Supplier No. 16R880
The Micro 100 Engraving Tool Single End 16R880 is a carbide engraving tool with a single end cutting edge. It has a shank diameter of 1/8", an overall length of 2", and a cutting diameter of 0.040". This engraving tool is made of solid carbide and can machine a variety of materials including steel, aluminum, brass, copper, plastics, and wood. With its micro grain carbide construction, it provides excellent wear resistance and tool life. The single end cutting edge allows for detail engraving work in tight areas. This is a high precision engraving tool well-suited for detailed machining in applications like engraving, die sinking, cavity sinking, and micro machining.